Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MP Biomedicals™ Pepsin From Porcine Stomach Mucosa
Shop All MP Biomedicals, Inc Products
Click to view available options
Quantity:
50 g
500 g
Description
Pepsin contains the “cathepsin” component which has milk curdling activity. It has a broad range of substrate activity and demonstrates an esterase acitivity.
- CAS Number: 9001-75-6
- EC Number: 232-629-3
- MDL Number: MFCD00081840
- Format: off white lyophilized powder
- Specific activity: ≥1:10000
Used in preparation of Fab fragments from antibodies and to hydrolyze dry cervical samples in mice

Specifications
Specifications
For Use With (Application) | Molecular Biology, Proteomics |
Molecular Weight (g/mol) | ∽35000 |
Name | Pepsin |
pH Range | 2.5 to 4.5 |
Quantity | 50 g |
Source | Porcine |
Storage Requirements | 15° to 30°C |
Form | Powder |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction