Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Pepsinogen A Antibody (3D1C6), Alexa Fluor™ 488 , Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP306077AF488
Description
Pepsinogen A Monoclonal antibody specifically detects Pepsinogen A in Human samples. It is validated for ELISASpecifications
Pepsinogen A | |
Monoclonal | |
Alexa Fluor 488 | |
EC 3.4.23, EC 3.4.23.1, FLJ58952, FLJ77962, pepsin A, pepsinogen 4, group I (pepsinogen A), pepsinogen A4, PGA3, PGA5 | |
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human Pepsinogen A (Met 1-Ala 388). | |
0.1 mL | |
Primary | |
Human | |
Purified |
ELISA | |
3D1C6 | |
50mM Sodium Borate | |
Mouse | |
Protein A purified | |
RUO | |
643847 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction