Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Pepsinogen A Antibody (961902), Alexa Fluor™ 532, R&D Systems™

Rat Monoclonal Antibody
Supplier: Novus Biologicals MAB9350AF532
Description
Pepsinogen A Monoclonal specifically detects Pepsinogen A in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| Pepsinogen A | |
| Monoclonal | |
| Alexa Fluor 532 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| PGA4 | |
| Mouse myeloma cell line NS0-derived recombinant human Pepsinogen A4/PGA4, Ile16-Ala388, Accession No. P00790 | |
| 0.1 mL | |
| 643847.0 | |
| Store at 4°C in the dark. |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| 961902 | |
| Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin | |
| EC 3.4.23, EC 3.4.23.1, FLJ58952, FLJ77962, pepsin A, pepsinogen 4, group I (pepsinogen A), pepsinogen A4, PGA3, PGA5 | |
| Rat | |
| Protein A or G purified | |
| Primary | |
| Human | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction