Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Pepsinogen A4/PGA4 Antibody (03), Alexa Fluor™ 405, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP312534AF405
Description
Pepsinogen A4/PGA4 Monoclonal antibody specifically detects Pepsinogen A4/PGA4 in Human samples. It is validated for ELISASpecifications
Pepsinogen A4/PGA4 | |
Monoclonal | |
Alexa Fluor 405 | |
Mouse | |
Protein A purified | |
RUO | |
643847 | |
Store at 4°C in the dark. | |
IgG2a |
ELISA | |
03 | |
50mM Sodium Borate | |
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human Pepsinogen A4/PGA4 (Accession#: NP_001073276.1; Met1-Ala388). | |
100 μg | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction