Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Pepsinogen A4/PGA4 Rabbit anti-Human, Clone: 21, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP290168100UL
Description
Pepsinogen A4/PGA4 Monoclonal specifically detects Pepsinogen A4/PGA4 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| Pepsinogen A4/PGA4 | |
| Monoclonal | |
| Unconjugated | |
| 0.2 um filtered solution in PBS | |
| Recombinant Human Pepsinogen A4/PGA4 Protein | |
| 100 μg | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| 21 | |
| Immunohistochemistry, Immunohistochemistry-Paraffin 1:500-1:2000 | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| 643847 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction