Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Pepsinogen C/PGC/Progastricsin Antibody (90/11), Alexa Fluor™ 594, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB10062819AF594
Description
Pepsinogen C/PGC/Progastricsin Monoclonal specifically detects Pepsinogen C/PGC/Progastricsin in Human samples. It is validated for ELISA.Specifications
Pepsinogen C/PGC/Progastricsin | |
Monoclonal | |
Alexa Fluor 594 | |
50 mM sodium borate with 0.05% sodium azide | |
PGC | |
Human pepsinogen II | |
0.1 mL | |
Primary | |
NB100-62819 is specific for human pepsinogen II. Less than 1% reactivity has been observed with human pepsinogen I (isozymogens 2-5), porcine pepsinogen, gastrin I, somatostatin, peptide YY and bombesin. Affinity: 1 x 109 l/mol | |
Store at 4°C in the dark. |
ELISA | |
90/11 | |
ELISA | |
EC 3.4.23, EC 3.4.23.3, FLJ99563, gastricsin, PEPC, pepsin C, Pepsinogen C, pepsinogen group II, PGII, preprogastricsin, progastricsin (pepsinogen C) | |
Mouse | |
Protein A or G purified | |
Phospho Specific | |
5225.0 | |
Human | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction