Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pepstatin A, 98%, PROMO, Thermo Scientific Chemicals

Catalog No. J60237MBPM
Click to view available options
Quantity:
25 mg

Protease inhibitor that acts on pepsin, renin and cathepsin D

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 26305-03-3
Molecular Formula C34H63N5O9

Specifications

For Use With (Application) Protease inhibitor that acts on pepsin, renin and cathepsin D
Molecular Weight (g/mol) 685.9 g/mol
Purity 98%
Content And Storage 2°C to 8°C
Form Powder
Quantity 25 mg
Product Type Protease Inhibitor

RUO - Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.