Learn More
Thermo Scientific™ Pepstatin A Protease Inhibitor PROMO
Description
The individual reagents include AEBSF, aprotinin, bestatin, E64, leupeptin, pepstatin A, and PMSF. Although Thermo Scientific Halt™ Protease Inhibitor Cocktails provide convenient and optimized broad-spectrum protease inhibition for routine cell lysis and assay needs, certain applications require a more customized approach. By offering the individual high-quality reagents used to make our cocktail products, we provide you with the tools needed to apply a protease inhibitor individually, supplement a cocktail, or combine several components to make customized protease inhibitor cocktails for specialized applications.
Highlights:
- High-quality reagents in convenient and affordable package sizes
- Purchase individually to prepare customized protease inhibitor cocktails
- Supplement Halt Protease Inhibitor Single-Use Cocktails, whose formulations are fully disclosed, to achieve desired concentrations of specific component reagents
Specifications
Specifications
Product Type | Protease Inhibitor |
Content And Storage | Store between 2°C and 8°C |
Form | Lyophilized |
Product Line | Pierce™ |
Reagent Type | Protease Inhibitor |
Inhibitors | Protease Inhibitor |
Quantity | 25 mg |
For Research Use Only. Not for use in diagnostic procedures.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.