Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PEPT1 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501980938
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-0938 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-0938 Supplier Bioss Supplier No. BS10588R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

In mammalian small intestine, the proton-coupled peptide transporter (PEPT) is responsible for the absorption of small peptides arising from digestion of dietary proteins. PEPT1, a hydrogen ion/peptide cotransporter, transports dipeptides and tripeptides, but not free amino acids or peptides with more than three amino acid residues. Its driving force for uphill transport requires proton binding and the presence of an inside-negative membrane potential. PEPT1 is 708 amino acid protein that contains 12 putative membrane-spanning domains. PEPT1 is expressed in Caco-2 cells. PEPT1 seems to play important roles in nutritional and pharmacological therapies. The mammalian kidney expresses a proton-coupled peptide transporter, PEPT2, that is responsible for the absorption of small peptides, as well as beta-lactam antibiotics and other peptide-like drugs, from the tubular filtrate.

Specifications

Antigen PEPT1
Applications Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene SLC15A1
Gene Accession No. P46059
Gene Alias Caco-2 oligopeptide transporter; D630032F02; HPECT1; HPEPT1; Intestinal H(+)/peptide cotransporter; intestinal H+/peptide cotransporter; intestinal peptide transporter PEPT1; macrophage oligopeptide transporter PEPT1; Oligopeptide transporter, small intestine isoform; PECT1; Pept1; Peptide transporter 1; pineal gland-specific PEPT1; Proton-coupled dipeptide cotransporter; SLC15A1; solute carrier family 15 (oligopeptide transporter), member 1; Solute carrier family 15 member 1
Gene Symbols SLC15A1
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human PEPT1.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6564
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.