Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Peptide YY Antibody (1), Alexa Fluor™ 350, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP105167AF350
Description
Peptide YY Monoclonal specifically detects Peptide YY in Human samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
Peptide YY | |
Monoclonal | |
Alexa Fluor 350 | |
50 mM sodium borate with 0.05% sodium azide | |
PYY | |
Synthetic human PYY adsorbed onto aluminum hydroxide gel | |
0.1 mL | |
Primary | |
Binds human PYY. Cross-reactivity with NPY or PP in solution is | |
Store at 4°C in the dark. |
ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
1 | |
ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin | |
Peptide tyrosine tyrosine, peptide YY, PYY1, PYY-I | |
Mouse | |
Protein A or G purified | |
Signal Transduction | |
5697.0 | |
Human | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction