Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Peptide YY Antibody (1) - BSA Free, Novus Biologicals™
SDP

Catalog No. NB018337 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
100 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB018337 0.025 mg
NBP105167 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB018337 Supplier Novus Biologicals Supplier No. NBP1051670.025MG
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Peptide YY Monoclonal specifically detects Peptide YY in Human samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, Sandwich ELISA, Sandwich ELISA Capture.

Specifications

Antigen Peptide YY
Applications Immunohistochemistry, ELISA, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 1
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution ELISA 1:16000, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500, Sandwich ELISA Capture
Formulation 10mM Phosphate (pH 7.4) and 0.5M NaCl with 0.05% Sodium Azide
Gene Alias Peptide tyrosine tyrosine, peptide YY, PYY1, PYY-I
Gene Symbols PYY
Host Species Mouse
Immunogen Synthetic human PYY adsorbed onto aluminum hydroxide gel
Purification Method Protein A or G purified
Quantity 0.025 mg
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 5697
Test Specificity Binds human PYY. Cross-reactivity with NPY or PP in solution is
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.