Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Peptide YY Antibody (4) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
$206.50 - $486.50
Specifications
Antigen | Peptide YY |
---|---|
Clone | 4 |
Concentration | 1 mg/mL |
Dilution | Western Blot 1:100-1:2000, ELISA 1:100-1:2000 |
Applications | ELISA, Western Blot |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB019346
![]() |
Novus Biologicals
NBP2235840.025ML |
0.025 mL |
Each for $206.50
|
|
|||||
NBP22358401
![]() |
Novus Biologicals
NBP2235840.1ML |
0.1 mL |
Each for $486.50
|
|
|||||
Description
Peptide YY Monoclonal specifically detects Peptide YY in Human samples. It is validated for Western Blot, ELISA.Specifications
Peptide YY | |
1 mg/mL | |
ELISA, Western Blot | |
Unconjugated | |
Mouse | |
Human | |
Peptide tyrosine tyrosine, peptide YY, PYY1, PYY-I | |
PYY | |
IgG1 κ | |
Protein A or G purified | |
Clone 4 binds human PYY(3-36) Based on inhibition ELISA, Clone 4 shows no cross-reaction with PYY(1-36), PYY from mouse, rat, dog or pig, or NPY. |
4 | |
Western Blot 1:100-1:2000, ELISA 1:100-1:2000 | |
Monoclonal | |
Purified | |
Signal Transduction | |
10mM Phosphate (pH 7.4) and 0.5M NaCl with 15mM Sodium Azide | |
5697 | |
Synthetic N-terminal fragment of human PYY(3-36) coupled to carrier | |
Primary | |
Store at 4C. Do not freeze. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title