Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Peptide YY Antibody (4), PerCP, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP223584PCP
Description
Peptide YY Monoclonal antibody specifically detects Peptide YY in Human samples. It is validated for Western Blot, ELISASpecifications
Peptide YY | |
Monoclonal | |
PerCP | |
PBS | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Human | |
Purified |
Western Blot, ELISA | |
4 | |
Western Blot, ELISA | |
Peptide tyrosine tyrosine, peptide YY, PYY1, PYY-I | |
Synthetic N-terminal fragment of human PYY(3-36) coupled to carrier. | |
0.1 mL | |
Signal Transduction | |
5697 | |
Store at 4C in the dark. | |
IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction