Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Peptidylarginine Deiminase Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA564014
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA564014 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA564014 Supplier Invitrogen™ Supplier No. PA564014
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: PSACLKLFQE KKEEGYGEAA QFDGLKHQAK RSINEMLADR HLQRDNLHAQ KCIDWN Highest antigen sequence identity to the following orthologs: Mouse - 82%, Rat - 82%.

This gene encodes a member of the peptidyl arginine deiminase family of enzymes, which catalyze the post-translational deimination of proteins by converting arginine residues into citrullines in the presence of calcium ions. The family members have distinct substrate specificities and tissue-specific expression patterns. The type I enzyme is involved in the late stages of epidermal differentiation, where it deiminates filaggrin and keratin K1, which maintains hydration of the stratum corneum, and hence the cutaneous barrier function. This enzyme may also play a role in hair follicle formation. This gene exists in a cluster with four other paralogous genes.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Peptidylarginine Deiminase
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.20 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene PADI1
Gene Accession No. Q9ULC6
Gene Alias HPAD10; hPAD-colony 10; PAD1; PADI1; PADI5; PDI; PDI1; PDI2; PDI3; PDI4; peptidyl arginine deiminase 1; peptidyl arginine deiminase, type I; peptidylarginine deiminase I; Protein-arginine deiminase type I; protein-arginine deiminase type-1
Gene Symbols PADI1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human Peptidylarginine Deiminase. Recombinant protein control fragment (Product #RP-103444).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 29943
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.