Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Peptidylarginine Deiminase Polyclonal Antibody

Rabbit Polyclonal Antibody

Supplier:  Invitrogen™ PA5119333

 View more versions of this product

Catalog No. PIPA5119333


Only null left
Add to Cart
This item is not returnable. View return policy

Description

Description

Peptidylarginine Deiminase Polyclonal Antibody for Western Blot, IHC (P)

This gene encodes a member of the peptidyl arginine deiminase family of enzymes, which catalyze the post-translational deimination of proteins by converting arginine residues into citrullines in the presence of calcium ions. The family members have distinct substrate specificities and tissue-specific expression patterns. The type I enzyme is involved in the late stages of epidermal differentiation, where it deiminates filaggrin and keratin K1, which maintains hydration of the stratum corneum, and hence the cutaneous barrier function. This enzyme may also play a role in hair follicle formation. This gene exists in a cluster with four other paralogous genes.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Peptidylarginine Deiminase
Polyclonal
Unconjugated
PADI1
HPAD10; hPAD-colony 10; PAD1; PADI1; PADI5; PDI; PDI1; PDI2; PDI3; PDI4; peptidyl arginine deiminase 1; peptidyl arginine deiminase, type I; peptidylarginine deiminase I; Protein-arginine deiminase type I; protein-arginine deiminase type-1
Rabbit
Antigen affinity chromatography, Protein A
RUO
18599, 29943
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunohistochemistry (Paraffin), Western Blot
0.5 mg/mL
PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4
Q9ULC6, Q9Z185
PADI1
Recombinant protein Peptidyl Arginine Deiminase Type I. The antigen corresponds to amino acid range 298-556 of the target protein.
200 μL
Primary
Human, Mouse
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.