Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Peptidylglycine alpha-Amidating Monooxygenase/PAM Rabbit anti-Mouse, Polyclonal, Novus Biologicals™
SDP

Catalog No. NB095301 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB095301 20 μL
NB095300 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB095301 Supplier Novus Biologicals Supplier No. NBP30301020UL
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptidylglycine alpha-Amidating Monooxygenase/PAM Polyclonal specifically detects Peptidylglycine alpha-Amidating Monooxygenase/PAM in Mouse samples. It is validated for Western Blot.

Specifications

Antigen Peptidylglycine alpha-Amidating Monooxygenase/PAM
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS with 50% glycerol, pH7.3.
Gene Alias PAL, pancreatic peptidylglycine alpha-amidating monooxygenase, peptidyl alpha-amidating enzyme, peptidyl-alpha-hydroxyglycine alpha-amidating lyase, peptidylamidoglycolate lyase, peptidylglycine 2-hydroxylase, peptidyl-glycine alpha-amidating monooxygenase, peptidylglycine alpha-amidating monooxygenase, peptidylglycine alpha-hydroxylating monooxygenase, PHM
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 800 to the C-terminus of human Peptidylglycine alpha-Amidating Monooxygenase/PAM (NP_620176.1).
Purification Method Affinity purified
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5066
Target Species Mouse
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.