Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PER1 Antibody, Novus Biologicals™
SDP

Catalog No. p-5079443 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP224589SS 0.025 mg
NBP224589 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP224589SS Supplier Novus Biologicals Supplier No. NBP224589SS
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 1 publication

PER1 Polyclonal specifically detects PER1 in Human, Mouse, Bovine, Canine, Equine, Primate samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunoblotting, Knockout Validated.

Specifications

Antigen PER1
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoblot, Immunoassay
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Western Blot 5-10 ug/ml, Immunohistochemistry, Immunohistochemistry-Paraffin 5 ug/ml, Immunoblotting reported in scientific literature (PMID 29937374), Knockout Validated reported in scientific literature (Hamilton et al)
Formulation 0.2 ml PBS and 0.05% BSA with 0.05% Sodium Azide
Gene Alias Circadian clock protein PERIOD 1, Circadian pacemaker protein Rigui, hPER1, KIAA0482, PERhPER, period (Drosophila) homolog 1, period circadian protein homolog 1, period homolog 1 (Drosophila), Period, drosophila, homolog of, RIGUIMGC88021
Gene Symbols PER1
Host Species Rabbit
Immunogen A portion of amino acids 1150-1200 of human PER1 was used as the immunogen.
Purification Method Affinity Purified
Quantity 0.025 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5187
Test Specificity The amino acid sequence used as immunogen is 100% homologous in human, horse, opossum, cattle, chimpanzee and dog, and 93% homologous in rat and mouse.
Target Species Human, Mouse, Bovine, Canine, Equine, Primate
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.