Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PER1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA585531
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA585531 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA585531 Supplier Invitrogen™ Supplier No. PA585531
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Keep as concentrated solution. Predicted reactivity: Mouse (84%), Rat (82%), Bovine (84%). Positive Control: HeLa, HeLa (10 uM MG132 treatment for 2 hr), 293T, PER1-transfected 293T. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

Circadian rhythmicity is a basic property of phylogenetically diverse organisms which range from animals and plants, to fungi. Regulation of endogenous biological clocks is regulated at the genetic level by a protein-mediated, autoregulatory feed-back loop. In mammals, several genes that encode members of the basic helix-loop helix (bHLH) PAS (PER-ARNT-SIM) transcription factor family have been shown to play a significant role in regulating circadian oscillations. Transactivation of CLOCK-induced genes is mediated via an E box enhancer (CACGTG) found upstream of target genes. CLOCK-ARNT3 heterodimers bind to E box regulatory elements and stimulate gene transcription. CLOCK has been shown to transactivate the mammalian homolog of Drosophila per. PER, in concert with the product of the mammalian timeless gene (TIM), negatively regulates its own transcription by blocking the activity of the CLOCK-BMAL1 transactivation complex.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PER1
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 20% glycerol and 0.025% ProClin 300; pH 7
Gene PER1
Gene Accession No. O15534
Gene Alias circadian clock protein PERIOD 1; Circadian pacemaker protein Rigui; hPER; hPER1; KIAA0482; mPer1; m-rigui; PER; PER1; per1 {ECO:0000312; period 1; period circadian clock 1; period circadian protein homolog 1; period homolog 1; Period, drosophila, homolog of; RGD:727863}; RIGUI; rPER1
Gene Symbols PER1
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human PER1. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5187
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.