Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PER2 Antibody - BSA Free, Novus Biologicals™
SDP

Catalog No. p-5079481 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB100125SS 0.025 mL
NB100125 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB100125SS Supplier Novus Biologicals Supplier No. NB100125SS
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 5 publications

PER2 Polyclonal specifically detects PER2 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin.

Specifications

Antigen PER2
Applications Western Blot, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1 - 2 ug/ml, Immunohistochemistry 1:200, Immunocytochemistry/Immunofluorescence 1:100-1:500, Immunoprecipitation reported in scientific literature (PMID 31390562), Immunohistochemistry-Paraffin 1:200. Use reported in scientific literature (PMID 31822134)
Formulation PBS with 0.02% Sodium Azide
Gene Accession No. O15055
Gene Alias Circadian clock protein PERIOD 2, FASPS, hPER2, KIAA0347period 2, period (Drosophila) homolog 2, period circadian protein 2, period circadian protein homolog 2, period homolog 2 (Drosophila)
Gene Symbols PER2
Host Species Rabbit
Immunogen Two peptides derived from human PER2 [UniProt# O15055]. Antibodies cross-reacting with each peptide were affinity purified against the appropriate peptide.
Purification Method Affinity Purified
Quantity 0.025 mL
Regulatory Status RUO
Research Discipline Cancer, Chromatin Research, Circadian Rhythm, Endothelial Cell Markers, Hypoxia, Lipid and Metabolism, Neuroscience, Signal Transduction, Transcription Factors and Regulators, Vision
Primary or Secondary Primary
Gene ID (Entrez) 8864
Test Specificity Human. Peptide 1 has 94% homology to rat. Peptide 2 has 94% homology to rat and mouse.
Target Species Human, Mouse, Rat
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.