Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PER2 Antibody, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody has been used in 1 publication

$194.00

Specifications

Antigen PER2
Concentration 0.5 mg/mL
Dilution Immunohistochemistry-Paraffin 5 ug/ml
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP224596SS
SDP
View Documents
Novus Biologicals
NBP224596SS
0.025 mg
Each for $194.00
Only null left
Add to Cart
 
NBP224596
SDP
View Documents
Novus Biologicals
NBP224596
0.1 mg N/A N/A N/A
Description

Description

PER2 Polyclonal specifically detects PER2 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications

Specifications

PER2
Immunohistochemistry-Paraffin 5 ug/ml
Polyclonal
Purified
RUO
Human
O15055
8864
A portion of amino acids 1010-1060 of human PER2 was used as the immunogen.
Primary
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
0.5 mg/mL
Immunohistochemistry (Paraffin)
Unconjugated
Rabbit
Cancer, Chromatin Research, Circadian Rhythm, Endothelial Cell Markers, Hypoxia, Lipid and Metabolism, Neuroscience, Signal Transduction, Transcription Factors and Regulators, Vision
0.2 ml PBS and 0.05% BSA with 0.05% Sodium Azide
Circadian clock protein PERIOD 2, FASPS, hPER2, KIAA0347period 2, period (Drosophila) homolog 2, period circadian protein 2, period circadian protein homolog 2, period homolog 2 (Drosophila)
PER2
IgG
Protein A purified
The amino acid sequence used as immunogen is 100% homologous in human, chimpanzee and rhesus monkey, 93% homologous in bovine, dog and horse, and 87% homologous in mouse and rat.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.