Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PER2 Recombinant Rabbit Monoclonal Antibody (2K9Y0)
GREENER_CHOICE

Catalog No. PIMA535644
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA535644 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA535644 Supplier Invitrogen™ Supplier No. MA535644
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

Immunogen sequence: MNGYAEFPPS PSNPTKEPVE PQPSQVPLQE DVDMSSGSSG HETNENCSTG RDSQGSDCDD SGKELGMLVE PPDARQSPDT FSLMMAKSEH NPSTSGCSSD.

PER2, also known as Period circadian protein homolog 2, is a mainly nuclear protein that shows nucleocytoplasmic shuttling which is effected by interaction with other circadian core oscillator proteins and/or by phosphorylation. PER2 belongs to the basic helix-loop-helix family of transcription factors and contains a PAC (PAS-associated C-terminal) domain and two PAS (PER-ARNT-SIM) domains. PER2 is a component of the circadian core oscillator, which includes the CRY proteins, CLOCK or NPAS2, BMAL1 or BMAL2, CSNK1D and/or CSNK1E, TIMELESS, and the PER proteins. This protein is thus essential for generating circadian rhythms and is a negative element in the circadian transcriptional loop which influences clock function by interacting with other circadian regulatory proteins and transporting them to the nucleus. Expression of PER2 is fairly wide spread with strong expression in skeletal muscle and pancreas and slight expression in lung. Defects in PER2 are a cause of familial advanced sleep-phase syndrome (FASPS).
TRUSTED_SUSTAINABILITY

Specifications

Antigen PER2
Applications ChIP Assay, ELISA, Western Blot
Classification Recombinant Monoclonal
Clone 2K9Y0
Concentration 0.6 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene Per2
Gene Accession No. O15055
Gene Alias circadian clock protein PERIOD 2; FASPS; FASPS1; hPER2; KIAA0347; mKIAA0347; mPer2; PER2; per2 {ECO:0000312; period 2; period circadian clock 2; period circadian protein 2; period circadian protein homolog 2; period circadian regulator 2; period homolog 2; RGD:61945}; rPER2
Gene Symbols Per2
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 1-100 of human PER2 (O15055).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8864
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.