Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PER3 Antibody (SR1135), Novus Biologicals™

Rabbit Monoclonal Antibody
$309.50 - $423.00
Specifications
Antigen | PER3 |
---|---|
Clone | SR1135 |
Dilution | Western Blot 1:500-1:2000 |
Applications | Western Blot |
Classification | Monoclonal |
Description
PER3 Monoclonal antibody specifically detects PER3 in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
PER3 | |
Western Blot 1:500-1:2000 | |
Monoclonal | |
Purified | |
RUO | |
Human, Mouse, Rat | |
Cell growth-inhibiting gene 13 protein, Circadian clock protein PERIOD 3, GIG13, growth-inhibiting protein 13, hPER3, period (Drosophila) homolog 3, period 3, period circadian protein 3, period circadian protein homolog 3, period homolog 3 (Drosophila) | |
A synthesized peptide derived from human PER3 (Uniprot #: P56645) | |
Primary | |
Store at -20° C. Avoid freeze/thaw cycles. |
SR1135 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Cardiovascular Biology, Endocrinology | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
8863 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title