Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PER3 Mouse anti-Human, Clone:5D5B8, Abnova™
Mouse monoclonal antibody raised against recombinant human PER3.
Supplier: Abnova Corporation MAB16605
Description
This gene is a member of the Period family of genes and is expressed in a circadian pattern in the suprachiasmatic nucleus, the primary circadian pacemaker in the mammalian brain. Genes in this family encode components of the circadian rhythms of locomotor activity, metabolism, and behavior. Circadian expression in the suprachiasmatic nucleus continues in constant darkness, and a shift in the light/dark cycle evokes a proportional shift of gene expression in the suprachiasmatic nucleus. The specific function of this gene is not yet known. [provided by RefSeq]Specifications
PER3 | |
Monoclonal | |
Unconjugated | |
ELISA (1:10000) Western Blot (1:500-1:2000) The optimal working dilution should be determined by the end user. |
|
PER3 | |
Mouse | |
100 μg | |
Human | |
Antibody | |
IgG1 |
ELISA, Western Blot | |
5D5B8 | |
period homolog 3 (Drosophila) | |
In PBS (0.05% sodium azide) | |
GIG13 | |
Recombinant protein corresponding to amino acid 723-954 of human PER3 from E. coli. | |
8863 | |
Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction