Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PER3 Recombinant Rabbit Monoclonal Antibody (9I3I6)
GREENER_CHOICE

Catalog No. PIMA538036
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA538036 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA538036 Supplier Invitrogen™ Supplier No. MA538036
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

Positive Samples: HeLa, Mouse lung, Mouse kidney, Rat lung Immunogen sequence: GRRGAKDEAL GEESGERWSP EFHLQRKLAD SSHSEQQDRN RVSEELIMVV QEMKKYFPSE RRNKPSTLDA LNYALRCVHS VQANSEFFQI L.

This gene is a member of the Period family of genes and is expressed in a circadian pattern in the suprachiasmatic nucleus, the primary circadian pacemaker in the mammalian brain. Genes in this family encode components of the circadian rhythms of locomotor activity, metabolism, and behavior. This gene is upregulated by CLOCK/ARNTL heterodimers but then represses this upregulation in a feedback loop using PER/CRY heterodimers to interact with CLOCK/ARNTL. Polymorphisms in this gene have been linked to sleep disorders. Multiple transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PER3
Applications ELISA, Western Blot
Classification Recombinant Monoclonal
Clone 9I3I6
Concentration 2 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol, 0.05% BSA and 0.02% sodium azide; pH 7.3
Gene PER3
Gene Accession No. O70361, P56645, Q8CJE2
Gene Alias 2810049O06Rik; Cell growth-inhibiting gene 13 protein; circadian clock protein PERIOD 3; FASPS3; GIG13; hPER3; mPER3; Per3; period circadian clock 3; period circadian protein 3; period circadian protein homolog 3; period homolog 3; period3; rper3
Gene Symbols PER3
Host Species Rabbit
Immunogen Recombinant protein (or fragment).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 18628, 78962, 8863
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.