Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Perchloric Acid, 0.100 N (N/10) in Glacial Acetic Acid, Ricca Chemical
SDP

Catalog No. 551532 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
500 mL
1 L
2.5 L
4 L
20 L
Packaging:
Amber Glass Bottle
Cubitainer
Safety-coated Glass Bottle
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
5515-32 1 L Amber Glass Bottle
5515-16 500 mL Amber Glass Bottle
5515-5PT 2.5 L Safety-coated Glass Bottle
5515-1 4 L Amber Glass Bottle
5515-5 20 L Cubitainer
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Catalog No. 5515-32 Supplier Ricca Chemical Company Supplier No. 551532
Add to Cart
Add to Cart
This item is not returnable. View return policy

Chemical Identifiers

CAS 64-19-7 , 108-24-7 , 7601-90-3 , 7732-18-5
Molecular Formula C2H4O2
Molecular Weight (g/mol) 60.05
MDL Number MFCD00036152
InChI Key QTBSBXVTEAMEQO-UHFFFAOYSA-N
IUPAC Name acetic acid
SMILES CC(O)=O

Specifications

Color Colorless to Yellow
Concentration 0.100N
CAS Min % 94.0, 2.33, 1.28
CAS Max % 98.0, 2.43, 1.33
Packaging Amber Glass Bottle
Solvent or Matrix Glacial Acetic Acid
Quantity 1 L
Physical Form Liquid
Synonym ethanoic acid, ethylic acid, acetic acid, glacial, methanecarboxylic acid, vinegar acid, glacial, acetasol, acide acetique, essigsaeure
PubChem CID 176
ChEBI CHEBI:15366
Grade Laboratory
Chemical Name or Material Perchloric Acid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.