Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Perchloric Acid, 0.100 Normal (N/10) in Acetic Acid, Reagents

Catalog No. 01334305
Click to view available options
Quantity:
500 mL
1 L
2.5 L
4 L
Packaging:
Glass Bottle
Safety Coated Glass

Perchloric Acid, 0.100 Normal (N/10) in Acetic Acid, Reagent, for general laboratory use. Manufactured in ISO 9001 facility. NIST Traceable.

Chemical Identifiers

CAS 64-19-7, 108-24-7, 7601-90-3
Molecular Formula C2H4O2
Molecular Weight (g/mol) 60.05
MDL Number MFCD00036152
InChI Key QTBSBXVTEAMEQO-UHFFFAOYSA-N
Synonym Chlorine Tetraoxide in Glacial Acetic Acid
IUPAC Name acetic acid
SMILES CC(O)=O

Specifications

pH <2
Color Colorless to Light Yellow
Boiling Point 118°C
Packaging Glass Bottle
Specific Gravity 1.0556
Quantity 2.5 L
Physical Form Liquid
Grade Reagent
Chemical Name or Material Perchloric Acid, 0.100 Normal (N/10) in Acetic Acid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.