Learn More
Perchloric Acid, 0.100 Normal (N/10) in P-Dioxane, Reagents
Specifications
Specifications
| Color | Colorless |
| Boiling Point | 103°C |
| Packaging | Glass Bottle |
| Specific Gravity | 1.0394 |
| Quantity | 500 mL |
| Physical Form | Liquid |
| Grade | Reagent |
| Chemical Name or Material | Perchloric Acid, 0.100 Normal (N/10) in P-Dioxane |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.