Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Perchloric Acid, 0.100 Normal (N/10) in P-Dioxane, Reagents

Catalog No. 01334310
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-334-310 1 L
01-334-309 4 L
01-334-308 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 01-334-310 Supplier Reagents Holdings Llc Supplier No. CP1202001C
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Perchloric Acid, 0.100 Normal (N/10) in P-Dioxane, Reagent, for general laboratory use. Manufactured in ISO 9001 facility. NIST Traceable.

Chemical Identifiers

CAS 123-91-1 , 7601-90-3 , 7732-18-5
Molecular Formula C4H8O2
Molecular Weight (g/mol) 88.11
InChI Key RYHBNJHYFVUHQT-UHFFFAOYSA-N
Synonym Chlorine Tetraoxide in 1,4-Dioxane
IUPAC Name 1,4-dioxane
SMILES C1COCCO1

Specifications

Color Colorless
Boiling Point 103°C
Packaging Glass Bottle
Specific Gravity 1.0394
Quantity 1 L
Physical Form Liquid
Grade Reagent
Chemical Name or Material Perchloric Acid, 0.100 Normal (N/10) in P-Dioxane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.