Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Perchloric Acid, 70% (Certified ACS), Fisher Chemical™
Shop All Fisherbrand Products
Click to view available options
Quantity:
1 lb.
8 lb.
Packaging:
Glass Bottle
Safe-Cote™ Bottle
Safe-Cote™ Glass
Specifications
Specifications
| pH | 0.1 |
| Color | Colorless |
| Melting Point | -18°C |
| Boiling Point | 203°C |
| Density | 1.660g/cm³ |
| Heavy Metals (as Pb) | 1ppm max. |
| Identification | Pass Test |
| Packaging | Glass Bottle |
| Vapor Pressure | 6.8mmHg at 25°C |
| Flash Point | 113°C |
| Show More |
Safety and Handling
| Hazard Statement | DANGER! |
| Precautionary Statement | Emergency Overview Oxidizer: Contact with combustible/organic material may cause fire. Use personal protective equipment. Keep away from clothing and other combustible materials. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Immediate medical attention is required. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Immediate medical attention is required. Move to fresh air. If breathing is difficult, give oxygen. Do not use mouth-to-mouth resuscitation if victim ingested or inhaled the substance; induce artifici. Immediate medical attention is required. Immediate medical attention is required. Do not induce vomiting. NFPA Health:3 Flammability:0 Instability:3 |
| DOTInformation | DOT Class 5.1, : Oxidizer |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction