Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Perchloric acid, ACS, 48-50%

Catalog No. AA44464AP
Click to view available options
Quantity:
500 mL
2.5 L

Maximum level of impurities: Color (APHA) 10, Residue after ignition 0.003%, Silicate and phosphate as SiO2 5ppm, Cl 0.001%, Nitrogen compounds (as N) 0.001%, SO4 0.001%, Heavy Metals (as Pb) 1ppm, Fe 1ppm | CAS: 7601-90-3 | ClHO4 | 100.45 g/mol

Perchloric acid is used as a precursor for ammonium perchlorate, which is used in rocket fuel. It is useful for etching of liquid crystal displays, etching of chrome, ore extraction and in analytical chemistry. It is used as a strong oxidizer and as a solvent for metals and alloys. It serves as a deproteinization reagent in food analysis. As a dehydrating agent, it is used for the determination of silica in iron and steel and in cement and other silicate materials. It is involved in the separation and determination of copper and other metals and alkali metals such as sodium and potassium. It is also used as an adjunct to increase the reduction potential of cerate salts in volumetric analysis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7601-90-3
Molecular Formula ClHO4
Molecular Weight (g/mol) 100.45
MDL Number MFCD00011325
InChI Key VLTRZXGMWDSKGL-UHFFFAOYSA-N
Synonym unii-v561v90bg2, perchloric acid, perchloric acid acid by mass forbidden, perchloric acid, acs reagent, perchlorsaeure, perchloric acid, ueberchlorsaeure, hyperchloric acid, hclo4
PubChem CID 24247
ChEBI CHEBI:29221
IUPAC Name perchloric acid
SMILES O[Cl](=O)(=O)=O

Specifications

Melting Point °C to 112°C
Density 1.41 g/cm3 at 20°C
Vapor Pressure 29mm Hg at 20°C
Assay Percent Range 48-50%
Quantity 500 mL
Physical Form Liquid
UN Number UN1802
Solubility Information Miscible with water.
Formula Weight 100.46
Grade ACS
Chemical Name or Material Perchloric acid
Show More Show Less
missing translation for 'healthHazard1' H271-H290-H302-H314-H335-H373
missing translation for 'healthHazard2' Keep away from combustible material. In case of contact with eyes, rinse immediately with plenty of water and seek medical advice. Wear suitable protective clothing, gloves and eye/face protection. In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible). This material and/or its container must be disposed of as hazardous waste.
missing translation for 'healthHazard3' P210-P220-P221-P234-P260-P264b-P270-P271-P280-P283-P304+P340-P305+P351+P338-P306+P360-P310-P330-P331-P353-P363-P370+P378q-P371+P380+P375-P390-P501c
DOTInformation DOT Class: 8, Packaging Group: II
EINECSNumber 231-512-4
RTECSNumber SC7500000
TSCA Yes

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.