Learn More
Perchloric Acid, ACS, 60-62%, Thermo Scientific Chemicals
Description
Perchloric acid is used as a precursor for ammonium perchlorate, which is used in rocket fuel. It is useful for etching of liquid crystal displays, etching of chrome, ore extraction and in analytical chemistry. It is used as a strong oxidizer and as a solvent for metals and alloys. It serves as a deproteinization reagent in food analysis. As a dehydrating agent, it is used for the determination of silica in iron and steel and in cement and other silicate materials. It is involved in the separation and determination of copper and other metals, alkali metals such as sodium and potassium. Also, used as an adjunct to increase the reduction potential of cerate salts in volumetric analysis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | °C to 12°C |
| Boiling Point | 203°C |
| Density | 1.54 |
| Flash Point | 113°C (235°F) |
| Quantity | 2 L |
| Physical Form | Liquid |
| Solubility Information | Miscible with water. |
| Formula Weight | 100.46 |
| Grade | ACS Reagent |
| Chemical Name or Material | Perchloric Acid |
Safety and Handling
| Hazard Category | H271-H290-H302-H314-H335-H373 |
| Precautionary Statement | P210-P220-P221-P234-P260-P264b-P270-P271-P280-P283-P304+P340-P305+P351+P338-P306+P360-P310-P330-P331-P353-P363-P370+P378q-P371+P380+P375-P390-P501c |
| DOTInformation | Hazard Class: 5.1; Packaging Group: I |
| EINECSNumber | 231-512-4 |
| RTECSNumber | SC7500000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.