Alfa Aesar™ Perchloric acid, Environmental Grade Plus, 65-71%

Manufacturer: Alfa Aesar™ 38750AP

Quantity 500mL

 View more versions of this product

Catalog No. AA38750AP

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 7601-90-3
    Formula Weight 100.46
    MDL Number MFCD00011325
    UN Number UN1873
    Merck Index 14,7153
    Chemical Name or Material Perchloric acid
    Grade Environmental Grade Plus

    Specifications

    Physical Form Liquid, Double Sub-Boiling Quartz Distillation
    Formula Weight 100.46
    Health Hazard 3 GHS P Statement
    P221-P283-P303+P361+P353-P305+P351+P338-P405-P501a
    Take any precaution to avoid mixing with combustibles.
    Wear fire/flame resistant/retardant clothing.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
    Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Boiling Point 203°C
    CAS 7601-90-3
    Health Hazard 2 GHS H Statement
    H271-H314
    May cause fire or explosion
    strong oxidizer.
    Causes severe skin burns and eye damage.
    RTECS Number SC7500000
    Quantity 500mL
    Flash Point 113° (235°F)
    Melting Point −12°C
    UN Number UN1873
    Merck Index 14,7153
    Density 1.67g/mL
    Health Hazard 1 Danger
    Color Colorless
    TSCA TSCA
    MDL Number MFCD00011325
    DOT Information Transport Hazard Class: 5.1; Packing Group: I; Proper Shipping Name: PERCHLORIC ACID
    Odor Odorless
    Chemical Name or Material Perchloric acid
    Assay 65 to 71%
    Grade Environmental Grade Plus

    GHS H Statement
    H271-H314
    May cause fire or explosion
    strong oxidizer.
    Causes severe skin burns and eye damage.

    GHS P Statement
    P221-P283-P303+P361+P353-P305+P351+P338-P405-P501a
    Take any precaution to avoid mixing with combustibles.
    Wear fire/flame resistant/retardant clothing.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
    Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit