Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Perchloric acid, Puriss. p.a., ACS Reagent, Reag. ISO, Reag. Ph. Eur., 70.0-72.0%, Honeywell Fluka™
Shop All Honeywell Research Chemicals Products
Click to view available options
Quantity:
500 mL
1 L
2.5 L
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured

Specifications
Specifications
Concentration | 70 to 72% |
Residue after Ignition | ≤0.003% (as SO4) |
Flash Point | 113°C (235.4°F) |
Quantity | 2.5 L |
Linear Formula | HClO4 |
UN Number | 1873 |
Formula Weight | 100.46g/mol |
Grade | ACS/puriss. p.a. |
Chemical Name or Material | Perchloric Acid, Reag. Ph. Eur. reag. ISO |
Safety and Handling
Hazard Statement | P210-P280-P303 + P361 + P353-P304 + P340 + P310-P305 + P351 + P338-P371 + P380 + P375 |
Precautionary Statement | H271-H290-H302-H314-H373 |
RTECSNumber | SC7500000 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction