Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Perchloric Acid, TraceSELECT™, 67 to 72%, Honeywell Fluka™
Click to view available options
Quantity:
100 mL
500 mL
1 L
Packaging:
Glass bottle
Description
TraceSELECT™ Ultra and TraceSELECT
- ultra-pure acids, bases, and salts for smelting and wet digestion in environmental, water, and food analysis
- sample preparation for trace analysis requires reagents of the highest purity; our TraceSELECT Ultra products for ultra-trace analysis at ppb and even ppt levels are produced by sub-boiling distillation
- the acids, bases, and salts in the TraceSELECT series have been developed for sample preparation and analysis in the ppb (μg/kg) trace range
- packaged in high-quality containers appropriate for each product
- the reagents are produced and bottled under clean-room conditions

Specifications
Specifications
pH | >1 |
Color | Colorless |
Melting Point | -18°C |
Boiling Point | 203°C |
Density | 1.66g/cm3 at 20°C |
Odor | Odorless |
Packaging | Glass bottle |
Quantity | 500 mL |
Physical Form | Liquid |
Linear Formula | HClO4 |
Show More |
Safety and Handling
Hazard Statement | P210-P280-P303 + P361 + P353-P304 + P340 + P310-P305 + P351 + P338-P371 + P380 + P375 |
DOTInformation | 8(5.1) / PGI |
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction