Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Perchloromethyl mercaptan, 97%

Catalog No. AC130030250
Click to view available options
Quantity:
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

CAS: 594-42-3 | CCl4S | 185.87 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 594-42-3
Molecular Formula CCl4S
Molecular Weight (g/mol) 185.87
MDL Number MFCD00037054
InChI Key RYFZYYUIAZYQLC-UHFFFAOYSA-N
Synonym perchloromethyl mercaptan, perchloromethylmercaptan, clairsit, trichloromethanesulfenyl chloride, methanesulfenyl chloride, trichloro, perchloromethanethiol, perchlormethylmerkaptan, trichloromethane sulfenyl chloride, trichloromethylsulfenyl chloride, trichloromethanesulphenyl chloride
PubChem CID 11666
IUPAC Name trichloromethyl thiohypochlorite
SMILES ClSC(Cl)(Cl)Cl

Specifications

Color Yellow
Density 1.7000g/mL
Boiling Point 146.0°C to 148.0°C
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Refractive Index 1.5370 to 1.5410
Linear Formula CCl3SCl
Quantity 25 g
Beilstein 03,135
Specific Gravity 1.7
Formula Weight 185.88
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material Perchloromethyl mercaptan, 97%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Toxic if swallowed.
Fatal if inhaled.
Harmful in contact with skin.
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre
EINECSNumber 209-840-4
RTECSNumber PB0370000
TSCA TSCA

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.