Learn More
Cytiva Percoll™ Centrifugation Media

Description
- Gradients can either be preformed or spontaneously generated by centrifugation at moderate speeds in an angle-head rotor
- Low viscosity of media allows cell isolations on preformed gradients to be completed in only a few minutes using low centrifugal forces (200 to 1000xG)
- Iso-osmotic gradients throughout cover a range of densities up to 1.3g/mL
- Nontoxic, almost chemically inert and does not adhere to membranes
- Adjustable to physiological ionic strength and pH; can be buffered within pH range 5.5 to 10.0 without change in properties
- Percoll is a silica sol with nondialyzable PVP coating; 15-30nm dia. bead size
For in vitro research use only.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications
Specifications
Quantity | 250 mL |
Content And Storage | 4–30°C |
For Use With (Application) | Cell Isolation |
Viscosity | 15cP |
Osmolality | < 25 mOsm/kg |
Color | Colorless to Slightly Yellow |
pH | 8.5 to 9.5 |
Product Type | Centrifugation Media |
Density | 1.135 g/mL |
Form | Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.