Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Honeywell™ Perfect Fit™ Aramid Fiber Seamless Knit Gloves
Click to view available options
Size:
2X-Large
Description
Gloves do not provide thermal heat protection
- Aramid fiber construction provides cut, slash and abrasion resistance
- Construction provides comfortable fit and durability
- No seams to rub or split
- Can be laundered to extend service life

Specifications
Specifications
ANSI Abrasion Rating | ANSI 1 |
Construction | Seamless Knit |
Coating Material | Uncoated |
Color | Yellow |
Cut Resistance Rating | ANSI A3 |
Material | Aramid Fiber |
Product Type | Cut-resistant Gloves |
Size | 2X-Large |
EN388 Standard | EN 388 |
For Use With (Application) | Automotive assembly, sheet metal, glass handling, metal stamping, metal fabrication, recycling, canning, injection moulding, paper/pulp industry |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction