Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Honeywell™ Perfect Fit™ CRT Cut-Resistant Gloves

Catalog No. p-7236140
Click to view available options
Size:
Small
Large
2X-Large
Glove Cuff Color:
Blue
White
Yellow
This item is not returnable. View return policy
This item is not returnable. View return policy

Perfect Fit CRT (Cut-Resistant Technology) gloves are a proprietary blend of cut-resistant gloves and provide ANSI Cut Level 4 protection.

  • Perfect Fit™ CRT (Cut-Resistant Technology) offers ANSI A4 cut protection
  • Lightweight, seamless knit
  • Proprietary blend of cut resistant fibers that offer 2 to 3 times cut protection of other similar weight aramid gloves
  • 13-gauge
  • Abrasion, cut and puncture resistance
TRUSTED_SUSTAINABILITY

Specifications

Construction Seamless Knit
Coating Material Uncoated
Color Black, Yellow
Cut Resistance Rating ANSI A4
Material Proprietary Cut-resistant Fiber
Product Type Cut-resistant Gloves
Size Small
For Use With (Application) Agriculture, manufacturing, municipal services, steel and metals, transportation
Gender Women's
Glove Cuff Color Blue
Glove Cuff Style Knit
Hem Color Blue
Model CRT13L
Series Perfect Fit™ CRT
Yarn Gauge 13
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.