Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Perfex™ Multipurpose Brushes

Catalog No. 19042034A Shop All Perfex Corp. Products
Click to view available options
Color:
Blue
Red
White

Requires

Requires 19-042-036 or 19-042-041 handle.

Provide exceptional scrubbing action on irregular surfaces as well as floors, walls, and ceilings with these moderately stiff poly brushes. Nonconductive polyester fibers are flared to maintain contact at any angle, even on uneven surfaces. Unlike nylon, the polyester fibers and block will not absorb bacteria, liquids, or odors. Ergonomically designed to eliminate awkward bending and reaching. Available in blue, white, and red.

Specifications

Product Type Multi-purpose Brush
Color Blue
For Use With (Application) Provides exceptional scrubbing action on a variety of irregular surfaces, as well as floors, walls and ceilings
For Use With (Equipment) Adjustable Polymer Handle
Material Polyester Fiber
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.