Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Perfluoro-1,2-dimethylcyclobutane, 97%, remainder 1,3-isomer, Thermo Scientific Chemicals

Catalog No. p-7046909
Click to view available options
Quantity:
25 g
100 g

Perfluoro-1,2-dimethylcyclobutane acts as a ligand for Suzuki coupling. It is involved in the infrared multiple-photon decomposition study of its decomposition products induced by the transverse excited atmospheric pressure pulsed carbon dioxide laser (TEA CO2laser) irradiation.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Perfluoro-1,2-dimethylcyclobutane acts as a ligand for Suzuki coupling. It is involved in the infrared multiple-photon decomposition study of its decomposition products induced by the transverse excited atmospheric pressure pulsed carbon dioxide laser (TEA CO2laser) irradiation.

Solubility
Immiscible with water.

Notes
Incompatible with strong oxidizing agents.strong acids and strong bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2994-71-0
Molecular Formula C6F12
Molecular Weight (g/mol) 300.05
MDL Number MFCD00013737
InChI Key RBTROQHBNLSUTL-UHFFFAOYNA-N
Synonym Dodecafluorodimethylcyclobutane
PubChem CID 12789874
SMILES FC(F)(F)C1(F)C(F)(F)C(F)(F)C1(F)C(F)(F)F

Specifications

Melting Point -32°C
Density 1.62
Boiling Point 45°C
Refractive Index 1.254
Quantity 25 g
Solubility Information Immiscible with water.
IUPAC Name (3R,4R)-1,1,2,2,3,4-hexafluoro-3,4-bis(trifluoromethyl)cyclobutane
Formula Weight 300.05
Percent Purity 97%
Assay remainder 1,3-isomer
Chemical Name or Material Perfluoro-1,2-dimethylcyclobutane
Show More Show Less
EINECSNumber 221-065-3
TSCA Yes

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.