Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Perfluoro(4-methyl-2-pentene), [(E):(Z) 9:1], 90+%, cont 5-10% perfluoro(2-methyl-2-pentene)

Catalog No. AAL1683614
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1683614 25 g
AAL1683622 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1683614 Supplier Thermo Scientific Chemicals Supplier No. L1683614
Add to Cart

CAS: 2070-70-4 | C6F12 | 300.05 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2070-70-4
Molecular Formula C6F12
Molecular Weight (g/mol) 300.05
MDL Number MFCD00153253
InChI Key SAPOZTRFWJZUFT-UHFFFAOYSA-N
Synonym 2z-1,1,1,2,3,4,5,5,5-nonafluoro-4-trifluoromethyl pent-2-ene
PubChem CID 11012007
SMILES FC(=C(F)C(F)(C(F)(F)F)C(F)(F)F)C(F)(F)F

Specifications

Refractive Index <1.3000
Linear Formula (CF3)2CFCF=CFCF3
Quantity 25 g
IUPAC Name 1,1,1,2,3,4,5,5,5-nonafluoro-4-(trifluoromethyl)pent-2-ene
Formula Weight 300.05
Percent Purity ≥90%
Assay contains 5 to 10% perfluoro(2-methyl-2-pentene)
Chemical Name or Material Perfluoro(4-methyl-2-pentene), [(E):(Z) 9:1]
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.