Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Perfluoro Acid Anhydrides (TFAA, PFAA and HFAA) Acylation Reagents

Catalog No. PI65192
Change view
Click to view available options
Standard Components:
Heptafluoro-butanoic acid anhydride
Pentafluoro-propanoic acid anhydride
Trifluoroacetic acid anhydride
Quantity:
25 g
100 g
10 x 1 mL
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Standard Components Quantity
PI65192 Pentafluoro-propanoic acid anhydride 25 g
PI63164 Heptafluoro-butanoic acid anhydride 10 x 1 mL
PI67363 Trifluoroacetic acid anhydride 100 g
PI63162 Heptafluoro-butanoic acid anhydride 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. PI65192 Supplier Thermo Scientific™ Supplier No. TS65192

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Prepare volatile and stable fluoracyl derivatives for GC/MS with the Thermo Scientific™ Perfluoro Acid Anhydrides (TFAA, PFAA and HFAA).

Prepare fluoracyl derivatives for GC/MS and for flame ionization and electron-capture detection techniques with the Thermo Scientific™ Perfluoro Acid Anhydrides (TFAA, PFAA and HFAA). These reagents react readily with alcohols, phenols, and amines.

  • Used to prepare fluoracyl derivatives for GC/MS
  • Produce stable volatile derivatives for FID and ECD techniques
TRUSTED_SUSTAINABILITY

Specifications

For Use With GC/MS
Quantity 25 g
Standard Components Pentafluoro-propanoic acid anhydride
Storage Room Temperature
Hazards Corrosive
Standard Concentration 98 to 100%

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.