Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Perfluoro-compound FC-3283(TM), Acros Organics

Click to view available options
Quantity:
100g
250g
316806-89-0
521
316806-89-0
521
Density 1.8000g/mL
Infrared Spectrum Authentic
Quantity 100g
Boiling Point 123.0°C to 133.0°C
Specific Gravity 1.8
Formula Weight 521
Chemical Name or Material Perfluoro-compound FC-3283&asterisk;