Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Perfluoro-tert-butanol, 99%

Click to view available options
Quantity:
5g
25g
2378-02-1
C4HF9O
236.04
MFCD00042092
XZNOAVNRSFURIR-UHFFFAOYSA-N
perfluoro-tert-butanol,nonafluoro-tert-butanol,perfluoro-tert-butyl alcohol,perfluoro-t-butanol,nonafluoro-tert-butyl alcohol,2-propanol, 1,1,1,3,3,3-hexafluoro-2-trifluoromethyl,nonafluor-terc.butanol,1,1,1,3,3,3-hexafluoro-2-trifluoromethyl propan-2-ol,1,1,1,3,3,3-hexafluoro-2-trifluoromethyl-2-propanol,nonafluor-terc.butanol czech
16924
1,1,1,3,3,3-hexafluoro-2-(trifluoromethyl)propan-2-ol
C(C(F)(F)F)(C(F)(F)F)(C(F)(F)F)O
2378-02-1
C4HF9O
236.04
MFCD00042092
XZNOAVNRSFURIR-UHFFFAOYSA-N
perfluoro-tert-butanol,nonafluoro-tert-butanol,perfluoro-tert-butyl alcohol,perfluoro-t-butanol,nonafluoro-tert-butyl alcohol,2-propanol, 1,1,1,3,3,3-hexafluoro-2-trifluoromethyl,nonafluor-terc.butanol,1,1,1,3,3,3-hexafluoro-2-trifluoromethyl propan-2-ol,1,1,1,3,3,3-hexafluoro-2-trifluoromethyl-2-propanol,nonafluor-terc.butanol czech
16924
1,1,1,3,3,3-hexafluoro-2-(trifluoromethyl)propan-2-ol
C(C(F)(F)F)(C(F)(F)F)(C(F)(F)F)O
Quantity 5g
Boiling Point 44°C to 45°C
Odor Characteristic
Melting Point -17°C
Linear Formula (CF3)3COH
UN Number UN2810
Beilstein 1841640
Formula Weight 236.04
Percent Purity 99%
Chemical Name or Material Perfluoro-tert-butanol

GHS H Statement
H331-H315-H319-H335
Toxic if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P260-P280f-P305+P351+P338-P309-P310
Do not breathe dust/fume/gas/mist/vapors/spray.
Wear protective clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.

RTECSNumber : UB6452700