Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chiron Perfluoroalkyl Ether Carboxylic Acid (PFEECAs) Standards
SDP

Catalog No. p-200087330 Shop All Spex CertiPrep Products
Click to view available options
Chemical Name or Material:
Perfluoro-2-methoxyacetic acid
Perfluoro-3,6-dioxaheptanoic acid
Perfluoro-3-methoxypropanoic acid
Perfluoro-4-methoxybutanoic acid

Catalog No. 15-129-350ME

Supplier: Chiron 15129.350ME

PFAS testing can be highly challenging which is why we make it easier with our convenient portfolio of PFAS reference materials. Advance environmental and health safety with standards you can trust.

  • Various singles available
  • Solutions in assorted solvents
  • Useful for various matrices
  • Custom standards of other PFAS compounds available on demand

Specifications

Chemical Name or Material Perfluoro-2-methoxyacetic acid
CAS 674-13-5
Concentration 50 μg/mL
Synonym PFMOAA
Type Reference Material
Solvent or Matrix Methanol
Quantity 1 mL
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.