Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Perfluorodecalin, 90%, mixture of cis and trans, Thermo Scientific Chemicals

Catalog No. AC130045000
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass Bottle

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 306-94-5
Molecular Formula C10F18
Molecular Weight (g/mol) 462.08
MDL Number MFCD00010626
InChI Key UWEYRJFJVCLAGH-UHFFFAOYSA-N
Synonym perfluorodecalin, perflunafene, octadecafluorodecahydronaphthalene, cis-perfluorodecalin, trans-perfluorodecalin, perfluorodecahydronaphthalene, naphthalene, octadecafluorodecahydro, f-dc, pp 5, perflunafenum latin
PubChem CID 9386
ChEBI CHEBI:38848
IUPAC Name 1,1,2,2,3,3,4,4,4a,5,5,6,6,7,7,8,8,8a-octadecafluoronaphthalene
SMILES C12(C(C(C(C(C1(F)F)(F)F)(F)F)(F)F)(C(C(C(C2(F)F)(F)F)(F)F)(F)F)F)F

Specifications

Melting Point -10.0°C
Color Colorless
Density 1.9000g/mL
Boiling Point 142.0°C
Flash Point 59°C
Infrared Spectrum Authentic
Assay Percent Range 88% min. (GC)
Packaging Glass Bottle
Refractive Index 1.3135 to 1.3155
Quantity 500 g
Merck Index 15,4215
Specific Gravity 1.9
Solubility Information Solubility in water: insoluble
Formula Weight 462.08
Percent Purity 90%
Physical Form Liquid
Chemical Name or Material Perfluorodecalin
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Flammable liquid and vapor.
Precautionary Statement GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Store in a well-ventilated place.
Keep container tightly closed.
EINECSNumber 206-192-4
RTECSNumber QJ3175000
TSCA TSCA

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.