Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Perfluorododecanoic acid, 96%, Thermo Scientific Chemicals

Catalog No. AAL1411306
Click to view available options
Quantity:
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 307-55-1
Molecular Formula C12HF23O2
Molecular Weight (g/mol) 614.101
MDL Number MFCD00198081
InChI Key CXGONMQFMIYUJR-UHFFFAOYSA-N
Synonym perfluorododecanoic acid, tricosafluorododecanoic acid, perfluorolauric acid, dodecanoic acid, tricosafluoro, unii-p6g1j7nm2d, p6g1j7nm2d, pfdoa, tricosafluorolauric acid, n-perfluorododecanoic acid, n-c11f23cooh
PubChem CID 67545
IUPAC Name 2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,11,11,12,12,12-tricosafluorododecanoic acid
SMILES C(=O)(C(C(C(C(C(C(C(C(C(C(C(F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)O

Specifications

Melting Point 108°C to 112°C
Boiling Point 245°C
Flash Point None
Linear Formula CF3(CF2)10CO2H
Quantity 5 g
UN Number UN3261
Beilstein 1811257
Formula Weight 614.1
Percent Purity 96%
Chemical Name or Material Perfluorododecanoic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber 206-203-2
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.