Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Perfluorohexanes, 98+%, Thermo Scientific Chemicals

Catalog No. AA1581222
Click to view available options
Quantity:
25 g
100 g
500 g

Perfluorohexanes is used as a coolant, photosensitizer in fluorous biphasic singlet oxygenation and an inert reaction medium. It is used in the electronic cooling liquid and insulator. It plays a vital role as a reaction medium, especially for photooxidation reactions. Further, it finds application as an ultrasound contrast agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 355-42-0
Molecular Formula C6F14
Molecular Weight (g/mol) 338.044
MDL Number MFCD00000437
InChI Key ZJIJAJXFLBMLCK-UHFFFAOYSA-N
Synonym perfluorohexane, perflexane, tetradecafluorohexane, perfluoro-n-hexane, flutec pp1, hexane, tetradecafluoro, n-tetradecafluorohexane, fluorinert fc72, perfluorohexanes, perflexane usan:inn
PubChem CID 9639
ChEBI CHEBI:39427
IUPAC Name 1,1,1,2,2,3,3,4,4,5,5,6,6,6-tetradecafluorohexane
SMILES C(C(C(C(F)(F)F)(F)F)(F)F)(C(C(F)(F)F)(F)F)(F)F

Specifications

Melting Point -4°C
Color Colorless
Density 1.699 g/mL
Boiling Point 58°C to 60°C
Flash Point None
Refractive Index 1.251
Linear Formula CF3(CF2)4CF3
Quantity 100 g
Beilstein 1802113
Solubility Information Immiscible with water.
Formula Weight 338.04
Percent Purity ≥98%
Physical Form Liquid
Chemical Name or Material Perfluorohexanes
Show More Show Less
Hazard Category Warning
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
EINECSNumber 206-585-0
RTECSNumber MO4310000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.