Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(Perfluorohexyl)ethylene 96.0+%, TCI America™
SDP

Catalog No. P1102100G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P1102100G 100 g
P11025G 5 g
P110225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. P1102100G Supplier TCI America Supplier No. P1102100G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 25291-17-2
Molecular Formula C8H3F13
Molecular Weight (g/mol) 346.091
MDL Number MFCD00039249
InChI Key FYQFWFHDPNXORA-UHFFFAOYSA-N
Synonym perfluorohexyl ethylene, 1h,1h,2h-perfluoro-1-octene, tridecafluorohexyl ethylene, 1-octene, 3,3,4,4,5,5,6,6,7,7,8,8,8-tridecafluoro, unii-j4kbi9s48d, 3,3,4,4,5,5,6,6,7,7,8,8,8-tridecafluoro-1-octene, j4kbi9s48d, perfluorohexylethylene, acmc-209gix, c6f13ch=ch2
PubChem CID 91384
IUPAC Name 3,3,4,4,5,5,6,6,7,7,8,8,8-tridecafluorooct-1-ene
SMILES C=CC(C(C(C(C(C(F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F

Specifications

Color Colorless
Boiling Point 103°C
Quantity 100 g
UN Number 1993
Formula Weight 346.09
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material (Perfluorohexyl)ethylene
EINECSNumber (2)-4067
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.