Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Perfluorononane, 99%, Thermo Scientific Chemicals

Catalog No. AAL1736306
Click to view available options
Quantity:
5 g
25 g

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 375-96-2
Molecular Formula C9F20
Molecular Weight (g/mol) 488.07
MDL Number MFCD00153254
InChI Key UVWPNDVAQBNQBG-UHFFFAOYSA-N
Synonym perfluorononane, eicosafluorononane, nonane, eicosafluoro, perfluorononan, icosafluorononane, perfluoro-n-nonane, acmc-20ajxy, 1,1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,9-icosakis fluoranyl nonane
PubChem CID 520856
IUPAC Name 1,1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,9-icosafluorononane
SMILES FC(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F

Specifications

Melting Point -16°C
Density 1.799
Boiling Point 125°C to 126°C
Refractive Index 1.276
Linear Formula CF3(CF2)7CF3
Quantity 5 g
Formula Weight 488.07
Percent Purity 99%
Chemical Name or Material Perfluorononane
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.